Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Somatostatin R3/SSTR3 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7018R100UG
This item is not returnable.
View return policy
Description
Somatostatin R3/SSTR3 Monoclonal specifically detects Somatostatin R3/SSTR3 in Human samples. It is validated for Flow Cytometry.Specifications
Somatostatin R3/SSTR3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P32745 | |
SSTR3 | |
NS0 mouse myeloma cell line transfected with human Somatostatin R3/SSTR3 Accession # P32745 | |
RUO | |
6753 | |
Human | |
IgG2a |
Flow Cytometry | |
576017 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
somatostatin receptor 3, SomatostatinR3, SS-3-R, SS3-R, SSR-28, SST3, SSTR3 | |
Mouse | |
100 μg | |
Primary | |
Detects human Somatostatin R3/SSTR3 | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction