Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Somatostatin R3/SSTR3 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7018N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7018N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7018N100 Supplier R&D Systems Supplier No. FAB7018N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Somatostatin R3/SSTR3 Monoclonal specifically detects Somatostatin R3/SSTR3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Somatostatin R3/SSTR3
Applications Flow Cytometry
Classification Monoclonal
Clone 576017
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P32745
Gene Alias somatostatin receptor 3, SomatostatinR3, SS-3-R, SS3-R, SSR-28, SST3, SSTR3
Gene Symbols SSTR3
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Somatostatin R3/SSTR3 Accession # P32745
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6753
Test Specificity Detects human Somatostatin R3/SSTR3
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.