Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Somatostatin R5/SSTR5 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4448T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4448T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4448T100 Supplier R&D Systems Supplier No. IC4448T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Somatostatin R5/SSTR5 Monoclonal specifically detects Somatostatin R5/SSTR5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Somatostatin R5/SSTR5
Applications Flow Cytometry
Classification Monoclonal
Clone 394401
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAK61266
Gene Alias somatostatin receptor 5, somatostatin receptor subtype 5, somatostatin receptor type 5, SomatostatinR5, SS5R, SS-5-R, SS5-R, SST5, SSTR5
Gene Symbols SSTR5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Somatostatin R5/SSTR5 Met1-Leu364 Accession # AAK61266
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6755
Test Specificity Detects human Somatostatin R5/SSTR5 in direct ELISAs. Stains Somatostatin R5/SSTR5 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.