Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Sonic Hedgehog/Shh Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $409.50
Specifications
Antigen | Sonic Hedgehog/Shh |
---|---|
Clone | 605022 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB109391SP
![]() |
R&D Systems
MAB109391SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1093911
![]() |
R&D Systems
MAB109391100 |
100 μg |
Each for $409.50
|
|
|||||
Description
Sonic Hedgehog/Shh Monoclonal antibody specifically detects Sonic Hedgehog/Shh in Human samples. It is validated for nanSpecifications
Sonic Hedgehog/Shh | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS | |
E. coli-derived human Sonic Hedgehog/Shh protein, Cys24-Gly197, Accession # NP_000184.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
605022 | |
Unconjugated | |
Mouse | |
Human | |
NP_000184.1 | |
6469 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title