Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SorCS1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3457SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3457SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3457SP Supplier R&D Systems Supplier No. AF3457SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

SorCS1 Polyclonal specifically detects SorCS1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen SorCS1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8WY21.3
Gene Alias FLJ41758, FLJ43475, FLJ44957, hSorCS, SORCS, SORCS receptor 1, sorCS1, sortilin-related VPS10 domain containing receptor 1, VPS10 domain receptor protein SORCS 1, VPS10 domain receptor SorCS, VPS10 domain-containing receptor SorCS1
Gene Symbols SORCS1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human SorCS1 Ser111-Ser1099 (Ser231Gly) Accession # Q8WY21.3
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114815
Test Specificity Detects human SorCS1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse SorCS1 is observed and less than 1% cross-reactivity with recombinant human (rh) SorCS2 and rhSorCS3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.