Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SORCS2 Partial ORF (NP_065828.1, 802 a.a. - 900 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101568
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system. [provided by RefSeq]

Sequence: VLRVLDQFQVMPLQFSKELDAYNPNTPEWREDVGLVVTRLLSKETSVPQELLVTVVKPGLPTLADLYVLLPPPRPTRKRSLSSDKRLAAIQQVLNAQKI

Specifications

Accession Number NP_065828.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57537
Molecular Weight (g/mol) 36.63kDa
Name SORCS2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen VLRVLDQFQVMPLQFSKELDAYNPNTPEWREDVGLVVTRLLSKETSVPQELLVTVVKPGLPTLADLYVLLPPPRPTRKRSLSSDKRLAAIQQVLNAQKI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name SORCS2
Gene Symbol SORCS2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.