Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Sortilin Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB31541R100UG
This item is not returnable.
View return policy
Description
Sortilin Monoclonal specifically detects Sortilin in Human samples. It is validated for Flow Cytometry.Specifications
Sortilin | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
100 kDa NT receptor, Glycoprotein 95, Gp95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, Ntr3, SORT1, sortilin, sortilin 1 | |
Mouse myeloma cell line NS0-derived recombinant human Sortilin Ser78-Asn755 Accession # Q99523 | |
100 μg | |
6272 | |
Human | |
Supernatant |
Flow Cytometry | |
334703 | |
Flow Cytometry 0.25-1μg/106 cells | |
Q99523 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Sortilin in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction