Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sortilin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3154SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3154SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3154SP Supplier R&D Systems Supplier No. AF3154SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

Sortilin Polyclonal specifically detects Sortilin in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction.

Specifications

Antigen Sortilin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99523
Gene Alias 100 kDa NT receptor, Glycoprotein 95, Gp95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, Ntr3, SORT1, sortilin, sortilin 1
Gene Symbols SORT1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Sortilin Ser78-Asn755 Accession # Q99523
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6272
Test Specificity Detects human and mouse Sortilin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.