Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SOST/Sclerostin Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF1406 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1406 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1406 Supplier R&D Systems Supplier No. AF1406

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

SOST/Sclerostin Polyclonal specifically detects SOST/Sclerostin in Human samples. It is validated for Flow Cytometry∣Immunocytochemistry∣Immunohistochemistry∣Western Blot.

Specifications

Antigen SOST/Sclerostin
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Gene Accession No. Q9BQB4
Gene Alias sclerostin, VBCHsclerosteosis
Gene Symbols SOST
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human SOST Gln24-Tyr213 Accession # Q9BQB4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50964
Test Specificity Detects human SOST in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse SOST is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.