Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SOST/Sclerostin Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF1406
This item is not returnable.
View return policy
Description
SOST/Sclerostin Polyclonal specifically detects SOST/Sclerostin in Human samples. It is validated for Western Blot.Specifications
SOST/Sclerostin | |
Polyclonal | |
Biotin | |
Q9BQB4 | |
SOST | |
Mouse myeloma cell line NS0-derived recombinant human SOST (R&D Systems, Catalog # 1406-ST) Gln24-Tyr213 Accession # Q9BQB4 | |
50 μg | |
Primary | |
Detects human SOST in Western blots. In this format, approximately 20% cross-reactivity with recombinant mouse SOST is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
sclerostin, VBCHsclerosteosis | |
Goat | |
Affinity Purified | |
RUO | |
50964 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction