Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SOX10 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC28642S100UG
This item is not returnable.
View return policy
Description
SOX10 Monoclonal specifically detects SOX10 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| SOX10 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| DOM, MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4, WS4C, WS4mouse, human homolog of | |
| E. coli-derived recombinant human SOX10, Met1-Ala118, Accession # P56693 | |
| 100 μg | |
| 6663 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2447A | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| P56693 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human SOX10 in direct ELISA. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction