Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SOX10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $351.50
Specifications
Antigen | SOX10 |
---|---|
Clone | 315808 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB28641SP
![]() |
R&D Systems
MAB28641SP |
25 μg |
Each for $200.50
|
|
|||||
MAB28641100
![]() |
R&D Systems
MAB28641100 |
100 μg |
Each for $351.50
|
|
|||||
Description
SOX10 Monoclonal specifically detects SOX10 in Human samples. It is validated for Immunocytochemistry.Specifications
SOX10 | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
DOM, MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4, WS4C, WS4mouse, human homolog of | |
E. coli-derived recombinant human SOX10, Met1-Ala118, Accession # P56693 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human SOX10 in direct ELISAs. |
315808 | |
Immunocytochemistry | |
Unconjugated | |
Human | |
P56693 | |
6663 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title