Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SOX10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.00
Specifications
| Antigen | SOX10 |
|---|---|
| Clone | 1017727 |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB28643SP
![]() |
R&D Systems
MAB28643SP |
25 μg |
Each for $200.50
|
|
|||||
MAB28643100
![]() |
R&D Systems
MAB28643100 |
100 μg |
Each for $444.00
|
|
|||||
Description
SOX10 Monoclonal specifically detects SOX10 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| SOX10 | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DOM, MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4, WS4C, WS4mouse, human homolog of | |
| Synthetic human SOX10 peptide, Accession # P56693 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1017727 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P56693 | |
| 6663 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title