Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SOX15 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4070
This item is not returnable.
View return policy
Description
SOX15 Polyclonal specifically detects SOX15 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
SOX15 | |
Polyclonal | |
Unconjugated | |
O60248 | |
SOX15 | |
E. coli-derived recombinant human SOX15 Met1-Leu233 Accession # O60248 | |
100 μg | |
Primary | |
Detects human SOX15 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) SOX3, rhSOX14, and rhSOX21 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
protein SOX-15, SOX20, SOX20Protein SOX-20, SOX26, SOX26SOX12, SOX27, SOX27Protein SOX-12, SRY (sex determining region Y)-box 15, SRY (sex determining region Y)-box 20 | |
Sheep | |
Affinity Purified | |
RUO | |
6665 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction