Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SOX17 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 22 publications
Supplier: R&D Systems MAB1924SP
This item is not returnable.
View return policy
Description
SOX17 Monoclonal specifically detects SOX17 in Human samples. It is validated for Immunocytochemistry.Specifications
SOX17 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q9H6I2 | |
SOX17 | |
E. coli-derived recombinant human SOX17 Asp177-Val414 Accession # Q9H6I2 | |
25 μg | |
Primary | |
Detects human SOX17 in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human SOX2, 3, 7, or 10. | |
Human | |
Purified |
Immunocytochemistry | |
245013 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64321 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction