Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SOX17 NorthernLights™ NL557-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. NL1924R
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NL1924R 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NL1924R Supplier R&D Systems Supplier No. NL1924R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SOX17 Polyclonal specifically detects SOX17 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen SOX17
Applications Immunocytochemistry
Classification Polyclonal
Conjugate NorthernLights 557 (NL557)
Dilution Immunocytochemistry
Gene Accession No. Q9H6I2
Gene Alias FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3
Gene Symbols SOX17
Host Species Goat
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64321
Test Specificity Detects human SOX17 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) SOX2, rhSOX6, and rhSOX15 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.