Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SOX7 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
$227.50 - $549.50
Specifications
Antigen | SOX7 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
SOX7 Polyclonal specifically detects SOX7 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SOX7 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
MGC10895, SOX7 transcription factor, SRY (sex determining region Y)-box 7, transcription factor SOX-7 | |
SOX7 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q9BT81 | |
83595 | |
E. coli-derived recombinant human SOX7 Ser249-Ala345 Accession # Q9BT81 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human SOX7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human SOX1, -2, -3, -9, -10, and -17 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title