Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SOX9 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF3075 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3075 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3075 Supplier Bio-techne Sales Corp Supplier No. AF3075
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 48 publications

SOX9 Polyclonal specifically detects SOX9 in Avian∣Chicken∣Human∣Monkey∣Mouse∣Rat∣Rhesus Macaque∣Transgenic Mouse∣Turtle∣Xenograft samples. It is validated for ChIP Assay∣Flow Cytometry∣Immunocytochemistry∣Immunocytochemistry/Immunofluorescence∣Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Simple Western∣Western Blot.

Specifications

Antigen SOX9
Applications ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P48436
Gene Alias campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9
Gene Symbols SOX9
Host Species Goat
Immunogen E. coli-derived recombinant human SOX9 Met1-Lys151 Accession # P48436
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6662
Test Specificity Detects human SOX9 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human SOX10 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Avian, Chicken, Human, Monkey, Mouse, Rat, Rhesus Macaque, Transgenic Mouse, Turtle, Xenograft
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.