Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human sP-Selectin/CD62P ELISA Kit
Click to view available options
Quantity:
6 Plates
Description
It contains recombinant human sP-Selectin and antibodies raised against the recombinant factor. It has been shown to accurately quantitate the recombinant sP-Selectin. Results obtained using natural human sP-Selectin showed linear curves that were parallel to the standard curves obtained using the recombinant kit standards. These results indicate that this kit can be used to determine relative mass values for natural human sP-Selectin
Specifications
Specifications
Assay Sensitivity | 0.5ng/mL |
Conjugate | HRP |
Cross Reactivity | Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
Sufficient For | 6 x 96-well plates |
Target Species | Human |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 6403 |
Quantity | 6 Plates |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction