Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Sp3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4256SP
This item is not returnable.
View return policy
Description
Sp3 Polyclonal specifically detects Sp3 in Human samples. It is validated for Western Blot.Specifications
Sp3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp686O1631, GC-binding transcription factor Sp3, Sp3 transcription factor, specificity protein 3, SPR2, SPR-2, transcription factor Sp3 | |
Goat | |
Affinity Purified | |
RUO | |
6670 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q02447 | |
SP3 | |
E. coli-derived recombinant human Sp3 Asp53-Ser309 Accession # Q02447 | |
25 μg | |
Primary | |
Detects human Sp3 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction