Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sp3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4256
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4256 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4256 Supplier R&D Systems Supplier No. AF4256
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Sp3 Polyclonal specifically detects Sp3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Sp3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q02447
Gene Alias DKFZp686O1631, GC-binding transcription factor Sp3, Sp3 transcription factor, specificity protein 3, SPR2, SPR-2, transcription factor Sp3
Gene Symbols SP3
Host Species Goat
Immunogen E. coli-derived recombinant human Sp3 Asp53-Ser309 Accession # Q02447
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6670
Test Specificity Detects human Sp3 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.