Learn More
Gibco™ Human SPARC Recombinant Protein, PeproTech®

Description
Recombinant Human SPARC/Osteonectin is a glycoprotein containing 286 amino acids that migrates at an apparent MW of 43.7 kDa by SDS-PAGE analysis due to the effect of glycosylation. The calculated molecular weight of Recombinant Human SPARC/Osteonectin is 32.7 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Specifications
Specifications
Accession Number | P09486 |
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 6678 |
Molecular Weight (g/mol) | 32.7 kDa |
Name | Human SPARC |
Quantity | 10 μg |
Source | CHO cells |
Regulatory Status | RUO |
Endotoxin Concentration | <1 EU/ μg |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.