Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sphingosine Kinase 1/SPHK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5536SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5536SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5536SP Supplier R&D Systems Supplier No. AF5536SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Sphingosine Kinase 1/SPHK1 Polyclonal specifically detects Sphingosine Kinase 1/SPHK1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen Sphingosine Kinase 1/SPHK1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_068807
Gene Alias EC 2.7.1.91, SK1, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1, SPK1
Gene Symbols SPHK1
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Sphingosine Kinase 1/SPHK1 Asp2-Leu398 Accession # NP_068807
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8877
Test Specificity Detects human SPHK1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human SPHK2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.