Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sphingosine Kinase 1/SPHK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB55361SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB55361SP 25 μg
MAB55361100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB55361SP Supplier R&D Systems Supplier No. MAB55361SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sphingosine Kinase 1/SPHK1 Monoclonal specifically detects Sphingosine Kinase 1/SPHK1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Sphingosine Kinase 1/SPHK1
Applications Immunocytochemistry
Classification Monoclonal
Clone 647705
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9NYA1
Gene Alias EC 2.7.1.91, SK1, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1, SPK1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Sphingosine Kinase 1/SPHK1, Asp2-Leu398, Accession No. Q9NYA1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 8877.0
Test Specificity Detects human Sphingosine Kinase 1/SPHK1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.