Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Spinesin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2495SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2495SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2495SP Supplier R&D Systems Supplier No. MAB2495SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Spinesin Monoclonal specifically detects Spinesin in Human samples. It is validated for Immunoprecipitation, Neutralization.

Specifications

Antigen Spinesin
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 376608
Conjugate Unconjugated
Dilution Immunoprecipitation 25 ug/mL, Neutralization 7 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q0P514
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, MGC141886, MGC148044, Spinesin, TMPRSS5, transmembrane protease serine 5, transmembrane protease, serine 5
Gene Symbols TMPRSS5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Spinesin Tyr27-Leu413 Accession # Q0P514
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80975
Test Specificity Detects human Spinesin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Spinesin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.