Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SPINK1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7496SP
This item is not returnable.
View return policy
Description
SPINK1 Polyclonal specifically detects SPINK1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
SPINK1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
pancreatic secretory trypsin inhibitor, PCTT, PCTTSpink3, PSTI, PSTISerine protease inhibitor Kazal-type 1, serine peptidase inhibitor, Kazal type 1, serine protease inhibitor, Kazal type 1, Spink3, TATI, TATITumor-associated trypsin inhibitor | |
Sheep | |
Affinity Purified | |
RUO | |
6690 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
P00995 | |
SPINK1 | |
E. coli-derived recombinant human SPINK1 Asp24-Cys79 Accession # P00995 | |
25 μg | |
Primary | |
Detects human SPINK1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction