Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SPIRE1 Full-length ORF (AAH16825.1, 1 a.a. - 131 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963082
Click to view available options
Quantity:
2 μg

Used for AP, Array, ELISA, WB-Re

Spire proteins, such as SPIRE1, are highly conserved between species. They belong to the family of Wiskott-Aldrich homology region-2 (WH2) proteins, which are involved in actin organization (Kerkhoff et al., 2001 [PubMed 11747823]).[supplied by OMIM]

Sequence: MRLPSKPYSTLPIFSLGPSALQRGESSMRSEKPSTAHHRPLRSIARFSSKSKSMDKSDEELQFPKELMEDWSTMEVCVDCKKFISEIISSSRRSLVLANKRARLKRKTQSFYMSSPGPSEYCPSERTISEI

Specifications

Accession Number AAH16825.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 56907
Molecular Weight (g/mol) 41.4kDa
Name SPIRE1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MRLPSKPYSTLPIFSLGPSALQRGESSMRSEKPSTAHHRPLRSIARFSSKSKSMDKSDEELQFPKELMEDWSTMEVCVDCKKFISEIISSSRRSLVLANKRARLKRKTQSFYMSSPGPSEYCPSERTISEI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC150621/MGC150622/Spir-1
Common Name SPIRE1
Gene Symbol SPIRE1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.