Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SPRED2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4819SP
This item is not returnable.
View return policy
Description
SPRED2 Polyclonal specifically detects SPRED2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
SPRED2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ31917, MGC163164, Spred-2related sequence, sprouty-related, EVH1 domain containing 2, sprouty-related, EVH1 domain-containing protein 2 | |
Sheep | |
Affinity Purified | |
RUO | |
200734 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q7Z698 | |
SPRED2 | |
E. coli-derived recombinant human SPRED2 Lys117-Ser297 Accession # Q7Z698 | |
25 μg | |
Primary | |
Detects human SPRED2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) SPRED1 is observed and less than 3% cross-reactivity with rhSPRED4 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction