Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SPRY1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6097SP
This item is not returnable.
View return policy
Description
SPRY1 Polyclonal specifically detects SPRY1 in Human samples. It is validated for Western Blot.Specifications
SPRY1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Drosophila, homolog of, 1 (antagonist of FGF signaling), sprouty homolog 1, antagonist of FGF signaling (Drosophila), spry-1 | |
Sheep | |
Affinity Purified | |
RUO | |
10252 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
O43609 | |
SPRY1 | |
E. coli-derived recombinant human SPRY1 Met1-Lys178 Accession # O43609 | |
25 μg | |
Primary | |
Detects human SPRY1 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction