Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SRCRB4D Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5654SP
This item is not returnable.
View return policy
Description
SRCRB4D Polyclonal specifically detects SRCRB4D in Human samples. It is validated for Western Blot.Specifications
SRCRB4D | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
S4D-SRCRB, S4-SRCRB, scavenger receptor cysteine rich domain containing, group B (4 domains), scavenger receptor cysteine-rich domain-containing group B protein, scavenger receptor cysteine-rich protein SRCRB-S4D, SRCRB-S4D, SSC4D | |
Sheep | |
Affinity Purified | |
RUO | |
136853 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q8WTU2 | |
SRCRB4D | |
Chinese hamster ovary cell line CHO-derived recombinant human SRCRB4D isoform 1 Ala45-Ser575 Accession # Q8WTU2 | |
25 μg | |
Primary | |
Detects human SRCRB4D in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human CD163 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction