Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SREBP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7119SP
This item is not returnable.
View return policy
Description
SREBP2 Monoclonal specifically detects SREBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
SREBP2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q12772 | |
SREBF2 | |
E. coli-derived recombinant human SREBP2 Leu242-Asp450 Accession # Q12772 | |
25 μg | |
Primary | |
Detects human SREBP2 in ELISA and Western Blot. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
751512 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBF2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6721 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction