Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SREC-I/SCARF1 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2409S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2409S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2409S100 Supplier R&D Systems Supplier No. FAB2409S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SREC-I/SCARF1 Monoclonal specifically detects SREC-I/SCARF1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SREC-I/SCARF1
Applications Flow Cytometry
Classification Monoclonal
Clone 373606
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q14162
Gene Alias Acetyl LDL receptor, KIAA0149endothelial cells scavenger receptor, MGC47738, SCARF1, scavenger receptor class F, member 1, Scavenger receptor expressed by endothelial cells 1, SREC1, SRECI, SREC-I, SRECscavenger receptor expressed by endothelial cells, SR-F1
Gene Symbols SCARF1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SREC-I/SCARF1 Ser20-Thr421 Accession # Q14162
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8578
Test Specificity Detects human SREC-I/SCARF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SREC-2 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.