Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SREC-II/SCARF2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2527
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2527 100 μg
AF2527SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2527 Supplier R&D Systems Supplier No. AF2527
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SREC-II/SCARF2 Polyclonal specifically detects SREC-II/SCARF2 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.

Specifications

Antigen SREC-II/SCARF2
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Gene Accession No. Q96GP6
Gene Alias HUMZD58C02, SCARF2, scavenger receptor class F, member 2, Scavenger receptor expressed by endothelial cells 2 protein, SREC2NSR1, SRECII, SREC-IIscavenger receptor class F member 2, SRECRP-1, SREPCR
Gene Symbols SCARF2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human SREC-II/SCARF2 Ala42-Gly442 Accession # Q96GP6
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91179
Test Specificity Detects SREC-II/SCARF2 in Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human SREC-I is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.