Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SSBP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6588
This item is not returnable.
View return policy
Description
SSBP1 Polyclonal specifically detects SSBP1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
SSBP1 | |
Polyclonal | |
Unconjugated | |
Q04837 | |
SSBP1 | |
E. coli-derived recombinant human SSBP1 Met1-Glu148 Accession # Q04837 | |
100 μg | |
Primary | |
Detects human SSBP1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
mitochondrial, MtSSB, single-stranded DNA binding protein 1 | |
Sheep | |
Affinity Purified | |
RUO | |
6742 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction