Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SSH1 Partial ORF (NP_061857.2, 950 a.a. - 1049 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963274
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

The ADF (actin-depolymerizing factor)/cofilin family (see MIM 601442) is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1; MIM 601329). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo (Niwa et al., 2002 [PubMed 11832213]).[supplied by OMIM]

Sequence: LVKQRTQEIETRLRLAGLTVSSPLKRSHSLAKLGSLTFSTEDLSSEADPSTVADSQDTTLSESSFLHEPQGTPRDPAATSKPSGKPAPENLKSPSWMSKS

Specifications

Accession Number NP_061857.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54434
Molecular Weight (g/mol) 36.74kDa
Name SSH1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen LVKQRTQEIETRLRLAGLTVSSPLKRSHSLAKLGSLTFSTEDLSSEADPSTVADSQDTTLSESSFLHEPQGTPRDPAATSKPSGKPAPENLKSPSWMSKS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ38102/KIAA1298/SSH-1
Common Name SSH1
Gene Symbol SSH1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.