Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ST2/IL-33R Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

$214.00 - $514.50

Specifications

Antigen ST2/IL-33R
Clone 2154D
Dilution Western Blot 2 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Conjugate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity & Availability  
MAB5233SP
GREENER_CHOICE
View Documents
R&D Systems
MAB5233SP
Unconjugated 25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB5233100
GREENER_CHOICE
View Documents
R&D Systems
MAB5233100
Unconjugated 100 μg
Each for $514.50
Only null left
Add to Cart
 
Description

Description

ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Human samples. It is validated for Western Blot.
Specifications

Specifications

ST2/IL-33R
Western Blot 2 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST-2/IL-33 R Lys19-Phe328 Accession # Q01638
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human ST-2/IL-33R in direct ELISAs.
2154D
Western Blot
Unconjugated
Rabbit
Human
Q01638
9173
IgG
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.