Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ST2/IL-33R Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $538.00
Specifications
| Antigen | ST2/IL-33R |
|---|---|
| Clone | 2154B |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5232SP
![]() |
R&D Systems
MAB5232SP |
25 μg |
Each for $214.00
|
|
|||||
MAB5232100
![]() |
R&D Systems
MAB5232100 |
100 μg |
Each for $538.00
|
|
|||||
Description
ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
| ST2/IL-33R | |
| Flow Cytometry 0.25 ug/10^6 cells, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST-2/ IL-33R Lys19-Phe328 Accession # Q01638 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ST-2/IL-33R in direct ELISAs. |
| 2154B | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q01638 | |
| 9173 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title