Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ST2/IL-33R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5231SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5231SP 25 μg
MAB5231100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5231SP Supplier R&D Systems Supplier No. MAB5231SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Human samples. It is validated for ELISA.

Specifications

Antigen ST2/IL-33R
Applications ELISA
Classification Monoclonal
Clone 97202
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. BAA02233
Gene Alias DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST2/IL-33 R Lys19-Phe328 Accession # BAA02233
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 9173
Test Specificity Detects human ST2/IL-33R in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.