Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ST2/IL-33R PE-conjugated Antibody, R&D Systems™

Catalog No. FAB5231P025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5231P025 25 Tests
FAB5231P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB5231P025 Supplier R&D Systems Supplier No. FAB5231P025
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

ST2/IL-33R Polyclonal specifically detects ST2/IL-33R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ST2/IL-33R
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. BAA02233
Gene Alias DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
Gene Symbols IL1RL1
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST2/IL-33 R Lys19-Phe328 Accession # BAA02233
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9173
Test Specificity Detects human ST2/IL-33 R in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-18 RI, and rhST-7 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.