Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ST6 Gal Sialyltransferase 1/ ST6GAL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5924
This item is not returnable.
View return policy
Description
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Monoclonal specifically detects ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 in Human samples. It is validated for Western Blot.Specifications
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P15907 | |
ST6GAL1 | |
Mouse myeloma cell line NS0-derived recombinant human ST6 Gal Sialytransferase 1/ST6GAL1 Lys27-Cys406 Accession # P15907 | |
100 μg | |
Primary | |
Detects human ST6 Gal Sialytransferase 1/ST6GAL1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ST6GAL2, rhST6GALNAC1, or rhST6GALNAC5 is observed. | |
Human | |
Purified |
Western Blot | |
396012 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Alpha 2,6-ST, B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CD75, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1, SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6480 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction