Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ST6 GalNAc alpha-2,6-sialyltransferase V/ST6GALNAC5 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$188.28 - $531.97

Specifications

Antigen ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5
Clone 719508
Concentration LYOPH
Dilution Immunocytochemistry 8-25 ug/mL
Applications Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB67151SP
GREENER_CHOICE
View Documents
R&D Systems
MAB67151SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
MAB67151
GREENER_CHOICE
View Documents
R&D Systems
MAB67151
100 μg
Each for $531.97
Only null left
Add to Cart
 
Description

Description

ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5 Monoclonal specifically detects ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5 in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5
LYOPH
Immunocytochemistry
Unconjugated
Mouse
Human
Q9BVH7
81849
Chinese hamster ovary cell line CHO-derived recombinant human SGalNAc alpha-2,6-sialyltransferase V/ST6GALNAC5 Gly30-Phe336 Accession # Q9BVH7
Primary
Sterile PBS to a final concentration of 0.5 mg/mL.
Detects human GalNAc alpha-2,6-sialyltransferase V/ST6GALNAC5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ST6GALNAC4 or rhST6GALNAC6 is observed.
719508
Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase E, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.7, GalNAc alpha-2,6-sialyltransferase V, GD1 alpha synthase, MGC3184, sialyltransferase 7 (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase) E, sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase) E, Sialyltransferase 7E, SIAT7E, SIAT7-E, SIAT7Ealpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase V, ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 5, ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 5, ST6GalNAc V, ST6GalNAcV, ST6GalNAcValpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
ST6GALNAC5
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.