Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ST7/LRP12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2560
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2560 100 μg
AF2560SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2560 Supplier R&D Systems Supplier No. AF2560
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ST7/LRP12 Polyclonal specifically detects ST7/LRP12 in Human samples. It is validated for Western Blot.

Specifications

Antigen ST7/LRP12
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9Y561
Gene Alias DKFZp762O2113, FAM4A1, LRP12, Protein FAM4A1, Protein HELG, RAY1, SEN4, subfamily A, member 1, suppression of tumorigenicity 7, suppression of tumorigenicity 7 (breast), suppressor of tumorigenicity 7 protein, TSG7
Gene Symbols LRP12
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human ST7/LRP12 Asn28-Ile488 Accession # Q9Y561
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29967
Test Specificity Detects human ST7/LRP12 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse ST7/LRP12 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.