Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6716SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6716SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6716SP Supplier R&D Systems Supplier No. AF6716SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q92185
Gene Alias Alpha-2,8-sialyltransferase 8A, alpha-N-acetylneuraminide alpha-2,8-sialyltransferase, disialoganglioside (GD3) synthase, Ganglioside GD3 synthase, Ganglioside GT3 synthase, ganglioside-specific alpha-2,8-polysialyltransferase, GD3 synthase), GD3S, sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3synthase) A, Sialyltransferase 8A, sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, Sialytransferase St8Sia I, SIAT8A, SIAT8-A, SIAT8EC 2.4.99.8, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1, ST8SiaI
Gene Symbols ST8SIA1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1 Tyr49-Ser356 Accession # Q92185
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6489
Test Specificity Detects human ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.