Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF6716
This item is not returnable.
View return policy
Description
ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 | |
Polyclonal | |
Unconjugated | |
Q92185 | |
ST8SIA1 | |
Chinese hamster ovary cell line CHO-derived recombinant human ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1 Tyr49-Ser356 Accession # Q92185 | |
100 μg | |
Primary | |
Detects human ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Alpha-2,8-sialyltransferase 8A, alpha-N-acetylneuraminide alpha-2,8-sialyltransferase, disialoganglioside (GD3) synthase, Ganglioside GD3 synthase, Ganglioside GT3 synthase, ganglioside-specific alpha-2,8-polysialyltransferase, GD3 synthase), GD3S, sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3synthase) A, Sialyltransferase 8A, sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, Sialytransferase St8Sia I, SIAT8A, SIAT8-A, SIAT8EC 2.4.99.8, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1, ST8SiaI | |
Sheep | |
Affinity Purified | |
RUO | |
6489 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction