Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ST8 alpha-2,8-Sialyltransferase 8B/ ST8SIA2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6590
This item is not returnable.
View return policy
Description
ST8 alpha-2,8-Sialyltransferase 8B/ST8SIA2 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 8B/ST8SIA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
ST8 alpha-2,8-Sialyltransferase 8B/ST8SIA2 | |
Polyclonal | |
Unconjugated | |
Q92186 | |
ST8SIA2 | |
Mouse myeloma cell line NS0-derived recombinant human alpha-2,8-Sialyltransferase 8B/ST8SIA2 Asp24-Thr375 Accession # Q92186 | |
100 μg | |
Primary | |
Detects human alpha-2,8-Sialyltransferase 8B/ST8SIA2 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.2 ug/mL, Immunohistochemistry 5-15 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
alpha-2,8-sialyltransferase 8B, alpha-2,8-sialyltransferase 8B 1, EC 2.4.99, EC 2.4.99.-, HsT19690, MGC116854, MGC116857, sialyltransferase 8 (alpha-2, 8-sialytransferase) B, Sialyltransferase 8B, Sialyltransferase X, Sialytransferase St8Sia II, SIAT8B, SIAT8-B, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2, ST8SiaII, ST8SIA-II, STX, STXST8SiaII | |
Sheep | |
Affinity Purified | |
RUO | |
8128 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction