Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Stabilin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3825SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3825SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3825SP Supplier R&D Systems Supplier No. AF3825SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Stabilin-1 Polyclonal specifically detects Stabilin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen Stabilin-1
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NY15
Gene Alias CLEVER-1, fasciclin egf-like, laminin-type egf-like, and link domain-containing scavengerreceptor-1, Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavengerreceptor 1, FEEL1, FEEL-1, FEEL-1MS-1 antigen, FELE-1, FEX1, KIAA0246common lymphatic endothelial and vascular endothelial receptor-1, MS-1 Antigen, STAB1, STAB-1, stabilin 1, stabilin-1, Stabilin1
Gene Symbols STAB1
Host Species Sheep
Immunogen E. coli-derived recombinant human Stabilin-1 Gly2208-Val2305 (Val2282Ile) Accession # Q9NY15
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23166
Test Specificity Detects human Stabilin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human Stabilin-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.