Learn More
Human Stabilin-1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Stabilin-1 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 840449 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q9NY15 |
| Gene Alias | CLEVER-1, fasciclin egf-like, laminin-type egf-like, and link domain-containing scavengerreceptor-1, Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavengerreceptor 1, FEEL1, FEEL-1, FEEL-1MS-1 antigen, FELE-1, FEX1, KIAA0246common lymphatic endothelial and vascular endothelial receptor-1, MS-1 Antigen, STAB1, STAB-1, stabilin 1, stabilin-1, Stabilin1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.