Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Stabilin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3645SP
This item is not returnable.
View return policy
Description
Stabilin-2 Monoclonal specifically detects Stabilin-2 in Human samples. It is validated for Immunohistochemistry.Specifications
Stabilin-2 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q8WWQ8 | |
STAB2 | |
E. coli-derived recombinant human Stabilin-2 Gly2198-Val2295 Accession # Q8WWQ8 | |
25 μg | |
Primary | |
Detects human Stabilin-2 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Stabilin-1 or recombinant mouse Stabilin-2 is observed. | |
Human | |
Purified |
Immunohistochemistry | |
841101 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD44-like precursor FELL, DKFZP434E0321, FAS1 EGF-like and X-link domain containing adhesion molecule-2, FAS1 EGF-like and X-link domain-containing adhesion molecule 2, fasciclin egf-like, laminin-type egf-like, and link domain-containing scavengerreceptor-2, Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavengerreceptor 2, FEEL2, FEEL-2, FELE-2, FELL, FELL-2, FEX2, HARE, HAREFELL2, hepatic hyaluronan clearance receptor, Hyaluronan receptor for endocytosis, hyaluronic acid receptor for endocytosis, STAB2, STAB-2, stabilin 2, stabilin-2, Stabilin2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
55576 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction