Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Stanniocalcin 2/STC-2 Protein
Click to view available options
Quantity:
50 μg
Description
The Recombinant Human Stanniocalcin 2/STC-2 Protein is derived from NS0. The Recombinant Human Stanniocalcin 2/STC-2 Protein has been validated for the following applications: Bioactivity.
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Accession Number | O76061 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μ filtered solution in PBS |
| Gene ID (Entrez) | 8614 |
| Molecular Weight (g/mol) | 31 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Stanniocalcin 2/STC-2 protein Thr25-Arg302,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction