Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human STAT1 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC1490S100UG
This item is not returnable.
View return policy
Description
STAT1 Monoclonal specifically detects STAT1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
STAT1 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P42224 | |
STAT1 | |
E. coli-derived recombinant human STAT1 Met1-Gln194 Accession # P42224 | |
RUO | |
6772 | |
Human | |
IgG2b |
Flow Cytometry | |
246523 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CANDF7, DKFZp686B04100, ISGF-3, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1, STAT91 | |
Rat | |
100 μg | |
Primary | |
Detects human STAT1. Detects recombinant human STAT1 transfectants but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction